missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (>99.0% n-Heptane), Honeywell Burdick & Jackson
$492.38 - $1446.10
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.21 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
IUPAC Name | heptane |
SMILES | CCCCCCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6001033
|
Honeywell-Burdick and Jackson
2071L |
6 x 1 L | Glass Bottle |
Case of 6 Each for $1,446.10
|
|
||||
6001032
|
Honeywell Inc
2074 |
4 x 4 L | Glass Bottle |
Each for $492.38
Case of 4 Each for $1,969.52
|
|
||||
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Specifications
142-82-5 | |
100.0000% | |
98.4°C | |
−4°C | |
C7H16 | |
UN1206 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
>99.9% | |
Heptane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (>99.0% n-Heptane) |
0.1 AU max. at 225 nm (UV), 0.01 AU max. at 250 nm (UV), 0.005 AU max. at 300 nm (UV) | |
0.684 g/cm3 | |
6 x 1 L | |
Glass Bottle | |
MFCD00009544 | |
1730763 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.21 | |
HPLC |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Heptanes