missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptafluorobutyric acid, For ion chromatography, ≥99.5% (GC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 5241125MLF
Specifications
375-22-4 | |
120°C (755 mmHg) | |
CF3 CF2 CF2 COOH | |
25 mL | |
7 (vs. air) | |
˜10 mmHg (25°C) | |
For ion chromatography |
1.645 g/mL (at 25°C) | |
Glass Bottle | |
MFCD00004171 | |
Edman Reagent No. 3; HFBA; Perfluorobutyric acid | |
214.04 | |
≥99.5% (GC) |
Chemical Identifiers
MFCD00004171 | |
Edman Reagent No. 3; HFBA; Perfluorobutyric acid |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
RTECSNumber : ET4025000