Learn More
Heidolph™ Tuttnauer™ PUREWISE™ Water Distiller Accessory
Carbon filter cartridge for PUREWISE Water Distiller
Supplier:  Heidolph™ 023212040CA
Catalog No. 13-876-808
Only null left
Explore available promotions
Description
Description
  • For use with Tuttnaeur Model 9000 (13-889-482)
  • Change after every 6 months of use
Specifications
Specifications
Water Distiller
Cartridge, Filter
Product Suggestions
Product Suggestions