missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Heating Blocks for Microtube Shaker
These nine interchangeable aluminum blocks accommodate PCR plates and a variety of common life science vessels.
Supplier: Axygen™ I4010C
Description
- No additional tools required for block exchanges
- Blocks utilize magnetic adhesion technology and are self-recognized once exchanged
- Combination of heating, cooling, and mixing with versatile block selection makes the Axygen™ Microtube Shaker ideal for many life science laboratories
- Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
Specifications
1500 rpm | |
Aluminum | |
Round Bottom |
2 mL | |
Axygen™ Microtube Shaker | |
24-well |