missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, heat inactivated, qualified, Canada, One Shot™ format DFS Item

Heat-inactivated serum in a One-Shot 50-mL bottle increases workflow benefits, while maintaining serum quality. The 50-mL bottle eliminates the need to aliquot and reduces the risk of contamination.

Supplier:  Gibco™ A3840302

 View more versions of this product

Catalog No. A3840302


Only null left
Explore available promotions

Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Serum category: Value (Standard)
    • For cell culture with robust cell lines
    • Good value for basic research
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
  • Origin: countries meeting USDA importation requirements

Gibco sera: trusted quality for consistent performance

  • ISO 13485-certified, processed in FDA-registered facilities
  • Triple filtered at 0.1 μm

Heat-inactivated and convenient One-Shot 50-mL bottle format

Heat-inactivated serum in a One-Shot 50-mL bottle increases workflow benefits, while maintaining serum quality. The 50-mL bottle eliminates the need to aliquot and reduces the risk of contamination. Purchasing heat-inactivated serum eliminates the multiple steps necessary to heat inactivate and improves product integrity and consistency since the heat inactivation is done in a controlled environment.

One-Shot 50-mL bottle features include:

  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label containing all of the information you need
  • 33% less waste generated than aliquoting from 500-mL bottles
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Fetal
One Shot
10 x 50 mL
Sterile
Qualified
Frozen
Bovine
Canada
Fetal Bovine Serum
Heat-inactivated
Triple-filtered, 0.1 μm
One Shot™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions