missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corepoint™ Scientific Hazardous Location Manual Defrost Freezer
Delivering professional protection for cold storage in hazardous laboratory locations.
Supplier: Corepoint™ Scientific EF141WWW0MHC
Description
Hazardous location freezers meet NFPA and OSHA guidelines for Hazardous Location, Class 1, Division II, Groups C and D guidelines.
- These units utilize nonsparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist.
- Models feature a microprocessor temperature controller, allowing for precise temperature control, recovery and maintenance versus mechanical style thermostats.
- Our freezers run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
- For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see Flammable Material Storage units.
Specifications
Explosion Proof Freezer | |
396 L | |
Solid Swing | |
Scratch-resistant Painted Steel | |
Cabinet foamed-in-place with EPA compliant high density urethane foam | |
White Powder Coated Steel | |
cULus, ETL, ENERGYSTAR | |
White | |
Two (2) Year Parts and Labor Warranty | |
EF141WWW/0MHC | |
3 Amps | |
115 V | |
Natural Hydrocarbon Refrigerant |
14 cu. ft. | |
-15°C to -25°C | |
1 | |
Painted Steel | |
Painted Steel | |
6 | |
Upright | |
Fan-forced | |
Manual | |
Microprocessor Temperature Controller | |
No Plug, Hardwired | |
H-drive |