Learn More
Hamilton™ 1000 Series Syringe, Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes the gas exchange in GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed.
Supplier: Merck Emd Millipore 20691
Specifications
Not Autoclavable | |
22 G | |
0.41 mm | |
0.72 mm | |
Gastight™ Syringe | |
122°F | |
50°F | |
2 | |
1 Ea. | |
Hamilton | |
Bevel |
Borosilicate Glass (Barrel), PTFE (Plunger), Stainless Steel (Tip) | |
0.0161 in. | |
0.0283 in. | |
2 in. | |
200 psig (13.8 bar) | |
50°C | |
1002 LTN | |
10°C | |
2.5 mL | |
51 mm | |
Luer Tip, Cemented |
GASTIGHT is a registered trademark of Hamilton Co.