Learn More
Honeywell Miller™ H500 Series Safety Harness, Model IC4
The modern ergonomic design provides high visibility and smart features, in addition to safe and comfortable functionality. Features highly breathable,lightweight, ergonomic shoulder/back padding.
Supplier:  Honeywell Miller™ H5IC221023
 View more versions of this product
Catalog No. 19-080-036
Only null left
Explore available promotions
Description
Description
  • Lightweight, ergonomic, breathable harness
  • Comfortable around the legs, waist, back and shoulders
  • Smart rebar hook/lanyard keepers and SRL web link
  • Increased freedom of movement
  • Quick and easy-to-fit and adjust
  • Improved visible stitch pattern
  • Clearly identifiable fall-impact indicators
Specifications
Specifications
Quick-Connect
2X-Large
Steel
Fall Protection Harness
H500 IC4 XXL STL QC SD
Side
Product Suggestions
Product Suggestions