Learn More
Honeywell Miller™ H500 Series Safety Harness, Model CC7
The modern ergonomic design provides high visibility and smart features, in addition to safe and comfortable functionality. Available in various models, each in three different sizes.
$170.97 - $170.97
Specifications
Buckle Style | Tongue/Mating |
---|---|
Material | Steel |
Product Type | Fall Protection Harness |
D-Ring Location | Side |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-080-010
|
Honeywell Miller™
H5CC311001 |
Small/Medium |
Each for $170.97
|
|
|||||
19-080-012
|
Honeywell Miller™
H5CC311003 |
2X-Large |
Each for $170.97
|
|
|||||
19-080-011
|
Honeywell Miller™
H5CC311002 |
Universal |
Each for $170.97
|
|
|||||
Description
- Lightweight, ergonomic, breathable harness
- Comfortable around the legs, waist, back and shoulders
- Smart rebar hook/lanyard keepers and SRL web link
- Increased freedom of movement
- Quick and easy-to-fit and adjust
- Improved visible stitch pattern
- Clearly identifiable fall-impact indicators
Specifications
Tongue/Mating | |
Steel | |
Fall Protection Harness | |
Side |