Learn More
Honeywell Miller™ H500 Series Safety Harness, Model CC5
The modern ergonomic design provides high visibility and smart features, in addition to safe and comfortable functionality. Available in various models, each in three different sizes.
Supplier: Honeywell Miller™ H5CC222023
Description
- Lightweight, ergonomic, breathable harness
- Comfortable around the legs, waist, back and shoulders
- Smart rebar hook/lanyard keepers and SRL web link
- Increased freedom of movement
- Quick and easy-to-fit and adjust
- Improved visible stitch pattern
- Clearly identifiable fall-impact indicators
Specifications
Quick-Connect | |
2X-Large | |
Aluminum |
Fall Protection Harness | |
H500 CC5 XXL ALUM QC SD | |
Side |