missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanfacine Hydrochloride 98.0+%, TCI America™
$115.57 - $734.44
Chemical Identifiers
CAS | 29110-48-3 |
---|---|
Molecular Formula | C9H10Cl3N3O |
Molecular Weight (g/mol) | 282.549 |
MDL Number | MFCD00798230 |
InChI Key | DGFYECXYGUIODH-UHFFFAOYSA-N |
Synonym | guanfacine hydrochloride, guanfacine hcl, tenex, estulic, intuniv, n-carbamimidoyl-2-2,6-dichlorophenyl acetamide hydrochloride, guanfacin hydrochloride, connexyn, n-amidino-2-2,6-dichlorophenyl acetamide hydrochloride |
PubChem CID | 71401 |
IUPAC Name | N-(diaminomethylidene)-2-(2,6-dichlorophenyl)acetamide;hydrochloride |
SMILES | C1=CC(=C(C(=C1)Cl)CC(=O)N=C(N)N)Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
G0414100MG
|
TCI America
G0414100MG |
100 mg |
Each for $115.57
|
|
|||||
G04141G
|
TCI America
G04141G |
1 g |
Each for $734.44
|
|
|||||
Chemical Identifiers
29110-48-3 | |
282.549 | |
DGFYECXYGUIODH-UHFFFAOYSA-N | |
71401 | |
C1=CC(=C(C(=C1)Cl)CC(=O)N=C(N)N)Cl.Cl |
C9H10Cl3N3O | |
MFCD00798230 | |
guanfacine hydrochloride, guanfacine hcl, tenex, estulic, intuniv, n-carbamimidoyl-2-2,6-dichlorophenyl acetamide hydrochloride, guanfacin hydrochloride, connexyn, n-amidino-2-2,6-dichlorophenyl acetamide hydrochloride | |
N-(diaminomethylidene)-2-(2,6-dichlorophenyl)acetamide;hydrochloride |
Specifications
29110-48-3 | |
White | |
MFCD00798230 | |
2811 | |
DGFYECXYGUIODH-UHFFFAOYSA-N | |
N-(diaminomethylidene)-2-(2,6-dichlorophenyl)acetamide;hydrochloride | |
71401 | |
≥98.0% (HPLC,T) | |
Guanfacine Hydrochloride |
213°C | |
C9H10Cl3N3O | |
100 mg | |
guanfacine hydrochloride, guanfacine hcl, tenex, estulic, intuniv, n-carbamimidoyl-2-2,6-dichlorophenyl acetamide hydrochloride, guanfacin hydrochloride, connexyn, n-amidino-2-2,6-dichlorophenyl acetamide hydrochloride | |
C1=CC(=C(C(=C1)Cl)CC(=O)N=C(N)N)Cl.Cl | |
282.549 | |
282.55 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : CY1481000
TSCA : No