missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanethidine sulfate, Thermo Scientific™
$191.00 - $586.20
Chemical Identifiers
CAS | 645-43-2 |
---|---|
Molecular Formula | C10H24N4O4S |
Molecular Weight (g/mol) | 296.39 |
MDL Number | MFCD00035403 |
InChI Key | YUFWAVFNITUSHI-UHFFFAOYSA-N |
IUPAC Name | N''-[2-(azocan-1-yl)ethyl]guanidine; sulfuric acid |
SMILES | OS(O)(=O)=O.NC(N)=NCCN1CCCCCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC468622500
|
Thermo Scientific Chemicals
468622500 |
250 mg |
Each for $191.00
|
|
|||||
AC468620010
|
Thermo Scientific Chemicals
468620010 |
1 g |
Each for $586.20
|
|
|||||
Chemical Identifiers
645-43-2 | |
296.39 | |
YUFWAVFNITUSHI-UHFFFAOYSA-N | |
OS(O)(=O)=O.NC(N)=NCCN1CCCCCCC1 |
C10H24N4O4S | |
MFCD00035403 | |
N''-[2-(azocan-1-yl)ethyl]guanidine; sulfuric acid |
Specifications
Glass Bottle | |
645-43-2 | |
Crystalline Powder/Crystals | |
97.5% to 102.5% | |
White to Light Beige | |
OS(O)(=O)=O.NC(N)=NCCN1CCCCCCC1 | |
296.39 | |
296.39 |
Available | |
Titration with HClO4 | |
C10H24N4O4S | |
MFCD00035403 | |
YUFWAVFNITUSHI-UHFFFAOYSA-N | |
N''-[2-(azocan-1-yl)ethyl]guanidine; sulfuric acid | |
250 mg | |
Guanethidine sulfate |
Safety and Handling
GHS07: Exclamation mark
ShelfLife : 2 years
EINECSNumber : 211-442-0
RTECSNumber : MF3150000
Recommended Storage : Normal conditions