Learn More
Synthware™ Graduated Borosilicate Glass Syringes
Graduated Borosilicate Glass Sample Syringes are constructed of borosilicate glass, and feature permanent graduations with a luer slip tip.
Supplier:  Synthware™ S371203
 View more versions of this product
Catalog No. 31502319
Only null left
Explore available promotions
Description
Description
  • Borosilicate glass sample syringes.
  • Permanent graduations.
  • Luer slip tip.
Specifications
Specifications
Borosilicate Glass (Barrel)
Yes
Reusable
5 mL
Leur Slip
Non-sterile
Precise Sampling
Syringe
10 Pk.
Removable Needle Syringe
Yes
Product Suggestions
Product Suggestions