missing translation for 'onlineSavingsMsg'
Learn More

Golgi protein 58k Monoclonal Antibody (K92024_4F3), Invitrogen™

Mouse Monoclonal Antibody

$1693.48 - $2847.38

Specifications

Antigen Golgi protein 58k
Clone K92024_4F3
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA556037
View Documents
Invitrogen™
MA556037
200 μg
Each for $2,847.38
Only null left
 
MA555657
View Documents
Invitrogen™
MA555657
50 μg
Each for $1,693.48
Only null left
 
Description

Description

Sequence of this protein is as follows: FSEGKNQEVI DAISGAITQT PGCVLLDVDA GPSTNRTVYT FVGPPECVVE GALNAARVAS RLIDMSRHQG EHPRMGALDV CPFIPVRGVS VDECVLCAQA FGQRLAEELD VPVYLYGEAA RMDSRRTLPA IRAGEYEALP KKLQQADWAP DFGPSSFVPS WGATATGARK FLIAFNINLL GTKEQAHRIA LNLREQGRGK DQPGRLKKVQ GIGWYLDEKN LAQVSTNLLD FEVTALHTVY EETCREAQEL SLPVVGSQLV GLVPLKALLD AAAFYCEKEN LFILEEEQRI RLVVSRLGLD SLCPFSPKER IIEYLVPERG

Golgi protein 58k Monoclonal antibody specifically detects Golgi protein 58k in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Specifications

Specifications

Golgi protein 58k
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide
58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1
Recombinant human FTCD (immunogen range is 11-330).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Ftcd
K92024_4F3
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Mouse
Human, Rat
O88618, O95954
10841, 89833
IgG1
Protein A
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'