missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Golgi protein 58k Monoclonal Antibody (K92024_1D11), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA555658

 View more versions of this product

Catalog No. MA555658


Only null left
Explore available promotions

Description

Description

Sequence of this protein is as follows: FSEGKNQEVI DAISGAITQT PGCVLLDVDA GPSTNRTVYT FVGPPECVVE GALNAARVAS RLIDMSRHQG EHPRMGALDV CPFIPVRGVS VDECVLCAQA FGQRLAEELD VPVYLYGEAA RMDSRRTLPA IRAGEYEALP KKLQQADWAP DFGPSSFVPS WGATATGARK FLIAFNINLL GTKEQAHRIA LNLREQGRGK DQPGRLKKVQ GIGWYLDEKN LAQVSTNLLD FEVTALHTVY EETCREAQEL SLPVVGSQLV GLVPLKALLD AAAFYCEKEN LFILEEEQRI RLVVSRLGLD SLCPFSPKER IIEYLVPERG

Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.
Specifications

Specifications

Golgi protein 58k
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
O88618, O95954
Ftcd
Recombinant human FTCD (immunogen range is 11-330).
50 μg
Primary
Human, Rat
Antibody
IgG1
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
K92024_1D11
Unconjugated
Ftcd
58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1
Mouse
Protein A
RUO
10841, 89833
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions