missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycoluril, 97%
CAS: 496-46-8 | C4H6N4O2 | 142.12 g/mol
$196.53 - $618.97
Chemical Identifiers
CAS | 496-46-8 |
---|---|
Molecular Formula | C4H6N4O2 |
Molecular Weight (g/mol) | 142.12 |
InChI Key | VPVSTMAPERLKKM-UHFFFAOYSA-N |
Synonym | glycoluril, acetyleneurea, acetylenediureine, acetylenediurea, glyoxalbiuret, glyoxaldiureine, acetylene carbamide, diurea glyoxalate, tetrahydroimidazo 4,5-d imidazole-2,5 1h,3h-dione, glyoxaldiurene |
PubChem CID | 62347 |
ChEBI | CHEBI:42946 |
IUPAC Name | 1,3,3a,4,6,6a-hexahydroimidazo[4,5-d]imidazole-2,5-dione |
SMILES | C12C(NC(=O)N1)NC(=O)N2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC120131000
|
Thermo Scientific Chemicals
120131000 |
100 g |
Each for $196.53
|
|
|||||
AC120135000
|
Thermo Scientific Chemicals
120135000 |
500 g |
Each for $618.97
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
496-46-8 | |
142.12 | |
glycoluril, acetyleneurea, acetylenediureine, acetylenediurea, glyoxalbiuret, glyoxaldiureine, acetylene carbamide, diurea glyoxalate, tetrahydroimidazo 4,5-d imidazole-2,5 1h,3h-dione, glyoxaldiurene | |
CHEBI:42946 | |
C12C(NC(=O)N1)NC(=O)N2 |
C4H6N4O2 | |
VPVSTMAPERLKKM-UHFFFAOYSA-N | |
62347 | |
1,3,3a,4,6,6a-hexahydroimidazo[4,5-d]imidazole-2,5-dione |
Specifications
496-46-8 | |
862.0°C | |
96% min. (N) | |
C4H6N4O2 | |
26,441 | |
glycoluril, acetyleneurea, acetylenediureine, acetylenediurea, glyoxalbiuret, glyoxaldiureine, acetylene carbamide, diurea glyoxalate, tetrahydroimidazo 4,5-d imidazole-2,5 1h,3h-dione, glyoxaldiurene | |
VPVSTMAPERLKKM-UHFFFAOYSA-N | |
1,3,3a,4,6,6a-hexahydroimidazo[4,5-d]imidazole-2,5-dione | |
62347 | |
142.12 | |
Glycoluril |
>300.0°C | |
Authentic | |
Plastic bottle | |
100 g | |
15,88 | |
Solubility in water: slightly soluble. Other solubilities: soluble in warm mineral acids and in warm ammonia | |
C12C(NC(=O)N1)NC(=O)N2 | |
142.12 | |
CHEBI:42946 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 207-821-5
RUO – Research Use Only