missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycolic Acid 98.0+%, TCI America™
$44.73 - $348.57
Chemical Identifiers
CAS | 79-14-1 |
---|---|
Molecular Formula | C2H4O3 |
Molecular Weight (g/mol) | 76.051 |
MDL Number | MFCD00004312 |
InChI Key | AEMRFAOFKBGASW-UHFFFAOYSA-N |
Synonym | glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech |
PubChem CID | 757 |
ChEBI | CHEBI:17497 |
IUPAC Name | 2-hydroxyacetic acid |
SMILES | C(C(=O)O)O |
Chemical Identifiers
79-14-1 | |
76.051 | |
AEMRFAOFKBGASW-UHFFFAOYSA-N | |
757 | |
2-hydroxyacetic acid |
C2H4O3 | |
MFCD00004312 | |
glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech | |
CHEBI:17497 | |
C(C(=O)O)O |
Specifications
0.1 (70% H2O soln. 25°C) | |
79-14-1 | |
C2H4O3 | |
3261 | |
AEMRFAOFKBGASW-UHFFFAOYSA-N | |
2-hydroxyacetic acid | |
757 | |
76.05 | |
25 g | |
Glycolic Acid |
76°C | |
White | |
MFCD00004312 | |
glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech | |
C(C(=O)O)O | |
76.051 | |
CHEBI:17497 | |
≥98.0% (GC,T) | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1346
RTECSNumber : MC5250000
TSCA : Yes