missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycolic Acid, 70%, High Purity, Spectrum™ Chemical
G1059, 79-14-1, C2H4O3
$307.00 - $44744.50
Chemical Identifiers
CAS | 79-14-1 |
---|---|
Molecular Formula | C2H4O3 |
Molecular Weight (g/mol) | 76.05 |
InChI Key | AEMRFAOFKBGASW-UHFFFAOYSA-N |
IUPAC Name | 2-hydroxyacetic acid |
SMILES | OCC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18603024
|
Spectrum Chemical Mfg Cor
G1059100ML |
100 mL | Amber Glass Bottle |
Each for $307.00
|
|
||||
18603026
|
Spectrum Chemical Mfg Cor
G1059500MLGL |
500 mL | Amber Glass Bottle |
Each for $748.77
|
|
||||
18603027
|
Spectrum Chemical Mfg Cor
G1059500MLGLCS6 |
6 x 500 mL | Amber Glass Bottle |
Case of 6 Each for $3,610.07
|
|
||||
18603028
|
Spectrum Chemical Mfg Cor
G10594LTGL |
4 L | Amber Glass Bottle |
Each for $2,157.50
|
|
||||
18603029
|
Spectrum Chemical Mfg Cor
G10594LTGLCS4 |
4 x 4 L | Amber Glass Bottle |
Case of 4 Each for $7,485.70
|
|
||||
18603025
|
Spectrum Chemical Mfg Cor
G105920LT |
20 L | Poly Drum |
Each for $5,912.30
|
|
||||
18603030
|
Spectrum Chemical Mfg Cor
G1059200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Glycolic Acid, 70 Percent, High Purity , is slightly stronger than acetic acid. It is often included into emulsion polymers, solvents and additives for ink and paint in order to improve flow properties and impart gloss. It is used in surface treatment products that increase the coefficient of friction on tile flooring. Glycolic acid is a useful synthetic organic intermediate for oxidation-reduction, esterification and long chain polymerizations.Chemical Identifiers
79-14-1 | |
76.05 | |
2-hydroxyacetic acid |
C2H4O3 | |
AEMRFAOFKBGASW-UHFFFAOYSA-N | |
OCC(O)=O |
Specifications
79-14-1,7732-18-5 | |
C2H4O3 | |
AEMRFAOFKBGASW-UHFFFAOYSA-N | |
2-hydroxyacetic acid | |
High Purity | |
Amber Glass Bottle |
70%,30% | |
UN3265 | |
OCC(O)=O | |
76.05 | |
100 mL |