missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, U.S.P. - F.C.C., J.T. Baker™
$1835.09 - $6034.41
Chemical Identifiers
CAS | 56-40-6 |
---|---|
Molecular Formula | C2H5NO2 |
Molecular Weight (g/mol) | 75.07 |
MDL Number | MFCD00008131 |
InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
Synonym | glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine |
PubChem CID | 750 |
ChEBI | CHEBI:15428 |
IUPAC Name | 2-aminoacetic acid |
SMILES | NCC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
02002945
|
Avantor J.T.Baker
058101 |
500 g | Wide Mouth Amber Glass Bottle |
Case of 4 Each for $1,835.09
|
|
||||
02002944
|
Avantor J.T.Baker
058105 |
2 kg | Wide Mouth Amber Glass Bottle |
Case of 4 Each for $6,034.41
|
|
||||
Chemical Identifiers
56-40-6 | |
75.07 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
750 | |
2-aminoacetic acid |
C2H5NO2 | |
MFCD00008131 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
CHEBI:15428 | |
NCC(O)=O |
Specifications
56-40-6 | |
Wide Mouth Amber Glass Bottle | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
NCC(O)=O | |
500 g | |
750 | |
75.07 | |
FCC/USP |
C2H5NO2 | |
MFCD00008131 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
2-aminoacetic acid | |
75.07 | |
CHEBI:15428 | |
1L = 1.16kg | |
Glycine |