Learn More

Glycerol (Clear, Colorless/USP/FCC), Fisher Chemical™
$317.69 - $663.00
Chemical Identifiers
CAS | 56-81-5 |
---|---|
Molecular Formula | C3H8O3 |
Molecular Weight (g/mol) | 92.09 |
MDL Number | MFCD00004722 |
InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
PubChem CID | 753 |
ChEBI | CHEBI:17754 |
IUPAC Name | propane-1,2,3-triol |
SMILES | OCC(O)CO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
G31-1
|
Thermo Fisher Scientific
G311 |
1 L | HDPE Bottle |
Each for $317.69
Case of 6 Each for $1,906.14
|
|
||||
G31-4
|
Thermo Fisher Scientific
G314 |
4 L | Glass Bottle |
Each for $663.00
Case of 4 Each for $2,652.00
|
|
||||
Chemical Identifiers
56-81-5 | |
92.09 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
753 | |
propane-1,2,3-triol |
C3H8O3 | |
MFCD00004722 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
CHEBI:17754 | |
OCC(O)CO |
Specifications
56-81-5 | |
18°C | |
5 | |
160°C | |
HDPE Bottle | |
HOCH2CH(OH)CH2OH | |
1 L | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
92.1 | |
95.0 to 100.5% (FCC) | |
Viscous Liquid |
>290°C | |
1.261g/cm³ | |
290°C | |
0.01% max. (FCC/USP) | |
C3H8O3 | |
MFCD00004722 | |
1.249 min. (FCC/USP) | |
Meets Requirements (USP) | |
OCC(O)CO | |
92.09 | |
CHEBI:17754 | |
0.003mbar at 50°C | |
FCC/USP | |
Glycerol, Clear, Colorless |
Safety and Handling

WARNING!
Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.
NFPA
Health:1
Flammability:1
Instability:0