missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8928 Class E Hi-Vis Insulated Bibs

GloWear™ 8928 Hi-Vis Insulated Bibs provide conspicuity, warmth, and durability for the toughest conditions.

$176.28 - $176.28

Specifications

Material Polyester
Color Yellow
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
View More Specs

Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-820-979
View Documents
Ergodyne™
25522
Small
Each for $176.28
Only null left
 
19-820-980
View Documents
Ergodyne™
25523
Medium
Each for $176.28
Only null left
 
19-820-981
View Documents
Ergodyne™
25524
Large
Each for $176.28
Only null left
 
19-820-982
View Documents
Ergodyne™
25525
X-Large
Each for $176.28
Only null left
 
19-820-983
View Documents
Ergodyne™
25526
2X-Large
Each for $176.28
Only null left
 
19-820-984
View Documents
Ergodyne™
25527
3X-Large
Each for $176.28
Only null left
 
19-820-985
View Documents
Ergodyne™
25528
4X-Large
Each for $176.28
Only null left
 
19-820-986
View Documents
Ergodyne™
25529
5X-Large
Each for $176.28
Only null left
 
Description

Description

  • ANSI Class E Complaint for low light conditions
  • Made with tough 300D polyester oxford to withstand wear and tear
  • PU transparent coating provides wind and water-resistant protection
  • Scuff-resistant knee patches for extra durability
  • Black accents in high-wear areas to hide dirt and grime
  • Adjustable straps and buckle release for comfort and fit
  • 1 zippered chest pocket and 2 side hand pockets for storage
  • Boot zippers for easy on and off without removing footwear
Specifications

Specifications

Polyester
Yellow
Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'