missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass Powder Funnels

Eisco™ Glass Powder Funnels designed with 60° angle stems are ideal for transferring powder reagents.

$11.53 - $39.60

Specifications

Material Borosilicate Glass
Autoclavable Autoclavable
Color Clear
Graduated No
For Use With (Application) Transferring powder
View More Specs

Products 6
Catalog Number Mfr. No. Diameter (Metric)Top Height (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric)Top Height (Metric) Price Quantity  
S89482
View Documents
Eisco™
CH0459A
45 mm 12.7 cm
Each for $11.53
Only null left
 
S89483
View Documents
Eisco™
CH0459B
55 mm 12.7 cm
Each for $11.53
Only null left
 
S89484
View Documents
Eisco™
CH0459C
75 mm 12.7 cm
Each for $16.50
Only null left
 
S89485
View Documents
Eisco™
CH0459D
85 mm 7.6 cm
Each for $19.00
Only null left
 
S89486
View Documents
Eisco™
CH0459E
100 mm 10.2 cm
Each for $20.87
Only null left
 
S89487
View Documents
Eisco™
CH0459F
120 mm 10.2 cm
Each for $39.60
Only null left
 
Description

Description

  • Made from high quality borosilicate 3.3 glass
  • Ideal for transferring powder reagents
  • 60° angle stem
  • Autoclavable

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Borosilicate Glass
Clear
Transferring powder
Filling Funnel
Autoclavable
No
1
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'