missing translation for 'onlineSavingsMsg'
Learn More

Glacial Acetic Acid, USP, 99.5-100.5%, Spectrum™ Chemical

AC110, 64-19-7, C2H4O2

Supplier:  Spectrum Chemical Mfg Cor AC11020LTPL

 View more versions of this product

Catalog No. 18602916


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Glacial Acetic Acid, USP does have some antibacterial and antifungal properties but is used primarily as a pH adjuster. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

64-19-7
C2H4O2
UN2789
CC(O)=O
60.05
USP
Poly Drum
100%
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
99.5 to 100.5%
20 L

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
CC(O)=O
C2H4O2
MFCD00036152
acetic acid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions