missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Girard's Reagent T 98.0+%, TCI America™
$52.19 - $350.44
Chemical Identifiers
CAS | 123-46-6 |
---|---|
Molecular Formula | C5H14ClN3O |
Molecular Weight (g/mol) | 167.64 |
MDL Number | MFCD00012009 |
InChI Key | YSULOORXQBDPCU-UHFFFAOYSA-N |
Synonym | girard's reagent t, girard reagent t, girard t reagent, girards reagent t, betaine hydrazide hydrochloride, unii-oo3mf0us46, trimethylaminoacetohydrazide chloride, carbazoylmethyltrimetylammonium chloride, trimethylacethydrazide ammonium chloride, trimethylammonium chloride acethydrazide |
PubChem CID | 67156 |
IUPAC Name | [(hydrazinecarbonyl)methyl]trimethylazanium chloride |
SMILES | [Cl-].C[N+](C)(C)CC(=O)NN |
Chemical Identifiers
123-46-6 | |
167.64 | |
YSULOORXQBDPCU-UHFFFAOYSA-N | |
67156 | |
[Cl-].C[N+](C)(C)CC(=O)NN |
C5H14ClN3O | |
MFCD00012009 | |
girard's reagent t, girard reagent t, girard t reagent, girards reagent t, betaine hydrazide hydrochloride, unii-oo3mf0us46, trimethylaminoacetohydrazide chloride, carbazoylmethyltrimetylammonium chloride, trimethylacethydrazide ammonium chloride, trimethylammonium chloride acethydrazide | |
[(hydrazinecarbonyl)methyl]trimethylazanium chloride |
Specifications
123-46-6 | |
White | |
MFCD00012009 | |
girard's reagent t, girard reagent t, girard t reagent, girards reagent t, betaine hydrazide hydrochloride, unii-oo3mf0us46, trimethylaminoacetohydrazide chloride, carbazoylmethyltrimetylammonium chloride, trimethylacethydrazide ammonium chloride, trimethylammonium chloride acethydrazide | |
[Cl-].C[N+](C)(C)CC(=O)NN | |
167.64 | |
167.64 | |
Crystalline Powder |
190°C | |
C5H14ClN3O | |
25 g | |
YSULOORXQBDPCU-UHFFFAOYSA-N | |
[(hydrazinecarbonyl)methyl]trimethylazanium chloride | |
67156 | |
≥98.0% (T) | |
Girard′s Reagent T |
Safety and Handling
EINECSNumber : (2)-3545
RTECSNumber : CP3150000
TSCA : Yes