missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM α, no nucleosides, powder DFS Item

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells.

$79.50 - $121.00

Specifications

Cell Line HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Cell Type Primary Rat Astrocytes
Classification Animal Origin-free
Form Powder
Product Type MEM α (Minimum Essential Medium α)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12000022 DFS Item
View Documents
Gibco™
12000022
10 x 1 L
Each for $121.00
Only null left
 
12000063 DFS Item
View Documents
Gibco™
12000063
10 L
Each for $79.50
Only null left
 
Description

Description

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of Gibco™ MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM α is modified as follows:

With: Phenol Red, L-glutamine

Without: Ribonucleosides, Deoxyribonucleosides, Sodium Bicarbonate

The complete formulation is available.

Gibco™ MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, vitamin B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR negative cells. This product is made with Earle's salts.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH. Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Animal Origin-free
MEM α (Minimum Essential Medium α)
Sterile-filtered
No HEPES, No Ribonucleosides, No Deoxyribonucleosides, No Sodium Bicarbonate
Gibco™
Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Primary Rat Astrocytes
Powder
Standard Serum Supplementation
Low Glucose, Glutamine, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
Room Temperature
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.