missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gentamicin sulfate

Gentamicin sulfate, CAS # 1405-41-0, is an aminoglycoside mainly used to inhibit the growth of Gram-negative bacteria.

$81.40 - $777.18

Chemical Identifiers

CAS 1405-41-0
Synonym Garamycin, Gentamicin Sulfate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC455310010
View Documents
Thermo Scientific Chemicals
455310010
1 g
Each for $81.40
Only null left
 
AC455310050
View Documents
Thermo Scientific Chemicals
455310050
5 g
Each for $221.85
Only null left
 
AC455310250
View Documents
Thermo Scientific Chemicals
455310250
25 g
Each for $777.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Gentamicin sulfate is a bactericidal aminoglycoside antibiotic which shows activity against Gram-negative bacteria
  • It works by inhibiting protein biosynthesis through binding the bacterial ribosome 30S subunit

Applications

  • Gentamicin sulfate has been used in studies to determine wound healing due to its antibiotic and antifungal properties
  • It has laboratory research applications in the study of a number of clinical conditions, including actinomycotic infection, appendicitis, cystic fibrosis, osteomyelitis, pelvic inflammatory disease, plague, gonorrhea, and tularemia
Specifications

Chemical Identifiers

1405-41-0
Garamycin, Gentamicin Sulfate

Specifications

Gentamicin Sulfate
218.0°C to 237.0°C
15% max.
Garamycin, Gentamicin Sulfate
≥590 IU/mg (Gentamycin)
1405-41-0
Authentic
1 g
Solubility in water: soluble.
Gentamicin sulfate
Videos
Safety and Handling

Safety and Handling

May cause allergy or asthma symptoms or breathing difficulties if inhaled, May cause an allergic skin reaction

Respiratory sensitisation (category 1), Skin sensitisation (category 1)

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only