missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genlantis™ PrimaPure™ Preadipocytes
High-quality human preadipocytes from adult heart or subcutaneous tissue
$2554.23 - $2554.23
Specifications
For Use With (Application) | Study insulin-stimulated glucose transport, growth hormone enhanced lypolysis and obese gene expression |
---|---|
Media Recommendation | Human Preadipocyte Growth Medium |
Quantity | >500000 Cells |
Source | Human Adipose Tissue |
Sufficient For | 16 Population Doublings |
Description
Each lot functionally characterized
Same quality standard as the PrimaPure human cell products
Each lot tested negative for HIV, Hepatitis B C, mycoplasma, bacteria, yeast, and fungi
Performance guaranteed with Genlantis media and reagents
Human Preadipocytes (HPAd) are fibroblast like precursor cells derived from human adipose tissue.
Specifications
Study insulin-stimulated glucose transport, growth hormone enhanced lypolysis and obese gene expression | |
>500000 Cells | |
16 Population Doublings |
Human Preadipocyte Growth Medium | |
Human Adipose Tissue |