missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ PrimaPure™ Fibroblasts Kits DFS Item

High-quality human adult, fetal, and neonatal fibroblasts

$1467.73 - $2295.00

Specifications

Content And Storage 4° and -20°C
For Use With (Application) For in-vitro analysis of fibroblast growth, migration and collagen metabolism in wound healing
Includes One ampoule of cryopreserved HDF, 500mL of HDF cell growth medium, one subculture reagent kit (100mL each of HEPES buffered saline solution, trypsin/EDTA, and trypsin neutralizing solution)
Media Recommendation Fibroblast Growth Medium
Quantity 1 Kit
View More Specs

Products 3
Catalog Number Mfr. No. Cell Type Source Price Quantity  
Catalog Number Mfr. No. Cell Type Source Price Quantity  
PH10605AK DFS Item
View Documents
Genlantis™
PH10605AK
Adult HDF Human Adult Skin
Each for $1,467.73
Only null left
 
PH10605FK DFS Item
View Documents
Genlantis™
PH10605FK
Fetal HDF Human Fetal
Each for $2,295.00
Only null left
 
PH10605NK DFS Item
View Documents
Genlantis™
PH10605NK
Neonatal HDF Human Neonatal Foreskin
Each for $1,929.02
Only null left
 
Description

Description

Specifications

Specifications

4° and -20°C
One ampoule of cryopreserved HDF, 500mL of HDF cell growth medium, one subculture reagent kit (100mL each of HEPES buffered saline solution, trypsin/EDTA, and trypsin neutralizing solution)
1 Kit
For in-vitro analysis of fibroblast growth, migration and collagen metabolism in wound healing
Fibroblast Growth Medium
16 Population Doublings
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'