missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ PrimaPure™ Epidermal Keratinocytes and Kits DFS Item

High-quality primary adult, fetal, and neonatal HEK

$1498.23 - $2336.94

Specifications

Content And Storage 4° and -20°C
For Use With (Application) In-vitro model for dermal toxicology tests in screening new skin products and for developing dermal skin substitutes, for investigations of human epidermal development, differentiation, and cellular aspects of skin diseases
Source Human Neonatal Foreskin or Adult Skin
Sufficient For 16 Population Doublings
View More Specs

Products 6
Catalog Number Mfr. No. Cell Type Quantity Price Quantity  
Catalog Number Mfr. No. Cell Type Quantity Price Quantity  
PH10205A DFS Item
View Documents
Genlantis™
PH10205A
Adult HEK >500000 Cells
Each for $2,336.94
Only null left
 
PH10205AK DFS Item
View Documents
Genlantis™
PH10205AK
Adult HEK 1 Kit
Each for $1,990.02
Only null left
 
PH10205F DFS Item
View Documents
Genlantis™
PH10205F
Fetal HEK >500000 Cells
Each for $1,517.29
Only null left
 
PH10205FK DFS Item
View Documents
Genlantis™
PH10205FK
Fetal HEK 1 Kit
Each for $2,070.08
Only null left
 
PH10205N DFS Item
View Documents
Genlantis™
PH10205N
Neonatal HEK >500000 Cells
Each for $1,498.23
Only null left
 
PH10205NK DFS Item
View Documents
Genlantis™
PH10205NK
Neonatal HEK 1 Kit
Each for $2,051.02
Only null left
 
Description

Description

  • Each lot is functionally characterized
  • Single donors to allow variability comparisons
  • Each lot tested negative for HIV, Hepatitis B C, mycoplasma, bacteria, yeast, and fungi
  • Supports 16 population doublings

Specifications

Specifications

4° and -20°C
In-vitro model for dermal toxicology tests in screening new skin products and for developing dermal skin substitutes, for investigations of human epidermal development, differentiation, and cellular aspects of skin diseases
Human Neonatal Foreskin or Adult Skin
16 Population Doublings
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'