Learn More
Corepoint Scientific™ General Purpose Refrigerator/Freezer Combo with Auto Defrost
This general purpose refrigerator and freezer combo unit provides versatile cold storage for scientific and industrial settings.
Supplier:  Corepoint Scientific™ LRF201WWW0A
Catalog No. 22-651-471
Only null left
Explore available promotions
Description
Description
  • Mechanical thermostat
  • Interior lighting and shelving
  • 2 probe access ports
  • High density urethane foam insulation
  • Natural hydrocarbon refrigerant
Specifications
Specifications
5.15 cu. ft.
146 L
-15°C to -10°C
-15°C to +10°C
Swing
Solid
Bottom-mounted
Mechanical Thermostat
Scratch-resistant Painted Steel
Urethane
CSA
Mechanical Thermostat
White
No
Two Probe Acess Ports (1 Refrigerator, 1 Freezer)
3 A
115 V
15.35 cu. ft.
435 L
0°C to 10°C
±0.1°C
2
2 Adjustable, 1 Fixed (Refrigerator), 1 Fixed (Freezer)
Automatic
Natural Hydrocarbon Refrigerant
White Powder-coated
White
Upright
No
Fan-forced
71 in.
Two Year Parts and Labor, Three Year Compressor Parts
NEMA 5-15 P
Product Suggestions
Product Suggestions