missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Gas Cylinder Barricade Rack for Cryogenic Cylinders
Justrite™ Gas Cylinder Barricade Rack for Cryogenic Cylinders meets USGS high-hazard seismic zone requirements when properly mounted
$1487.56 - $3038.89
Catalog Number | Mfr. No. | Holds | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Holds | Price | Quantity | |||||
19-085-798
|
Justrite™
35184 |
1 Cylinder 33 in. dia. |
Each for $1,848.94
|
|
|||||
19-085-799
|
Justrite™
35186 |
1 Cylinder 22 in. dia. |
Each for $1,487.56
|
|
|||||
19-085-800
|
Justrite™
35188 |
2 Cylinders 22 in. dia. |
Each for $2,436.51
|
|
|||||
19-085-801
|
Justrite™
35190 |
2 Cylinders 33 in. dia. |
Each for $3,038.89
|
|
|||||
Description
- Recommended for indoor or outdoor cryogenic cylinder storage
- Surround cylinders with 2-in (51-mm) square steel structural tubing
- Continuously seam welded at all junction points
- Sealed with exterior grade powder-coat paint for long lasting strength, durability and dependability
- Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries
- Features two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit
- Pre-drilled 0.531-in (14-mm) diameter floor mounting holes included on lower tube structure
- For seismic installations, add Model 35350 S4-rated anchor bolts (not included)
Compliance
OSHA, CGA P1, NFPA55
Warranty
1 YEAR
Specifications
Steel | |
Black | |
1016 mm |