missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAP43 Monoclonal Antibody (K29024_1C6), Invitrogen™
Mouse Monoclonal Antibody
$1557.73 - $2745.56
Specifications
Antigen | GAP43 |
---|---|
Clone | K29024_1C6 |
Concentration | 1 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Description
Sequence of this protein is as follows: VEKNDDDQKI EQDGIKPEDK AHKAATKIQA SFRGHITRKK LKGEKKDDVQ AAEAEANKKD EAPVADGVEK KGEGTTTAEA APATGSKPDE PGKAGETPSE
GAP43 Monoclonal antibody specifically detects GAP43 in Human, Rat samples. It is validated for Immunoprecipitation, Western BlotSpecifications
GAP43 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide | |
Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 | |
Recombinant human GAP43 (immunogen range is 11-110). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
GAP43 |
K29024_1C6 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
P07936, P17677 | |
2596, 29423 | |
IgG2b | |
Protein A | |
Antibody |