missing translation for 'onlineSavingsMsg'
Learn More

Gallic Acid, Purified, Spectrum™ Chemical

$679.77 - $6025.43

Chemical Identifiers

CAS 5995-86-8
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610691
View Documents
Spectrum Chemical Mfg Cor
G1004500GM
500 g Poly Bottle
Each for $679.77
Only null left
 
18610689
View Documents
Spectrum Chemical Mfg Cor
G100425KG
2.5 kg Poly Pail
Each for $2,893.60
Only null left
 
18610688
View Documents
Spectrum Chemical Mfg Cor
G100412KG
12 kg Fiber Drum
Each for $3,329.77
Only null left
 
18610690
View Documents
Spectrum Chemical Mfg Cor
G100425KGBL
25 kg Fiber Drum
Each for $6,025.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5995-86-8

Specifications

5995-86-8
Purified
Poly Bottle
1
500 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.