Learn More
Blackline Safety™ G7 Bridge, North America
Connects G7x person-worn devices with the Iridium satellite network.
Supplier:  Blackline Safety™ BRIDGENA
Catalog No. 170001972
Only null left
Explore available promotions
Description
Description
The G7 Bridge is self-powered, portable, and connects with up to five G7x devices from up to 1.2 miles (2 km) away.
Specifications
Specifications
G7 Bridge
Product Suggestions
Product Suggestions