missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vileda Professional™ Roll-O-Matic™ Self-Wringing Mops
Suitable for all controlled environments.
Supplier: Vileda Professional™ FBG120848
Description
- Mop with built-in wringer
- Wringer built into handle, eliminating need for expensive and bulky wringer
- Improved components eliminate sharp edges and provide for excellent refill retention
- Fits all original and advanced technology Roll-O-Matic approved refills
- Ergonomic grip
Specifications
58.5 to 76.25 x 10 in. | |
Blue | |
Not Autoclavable | |
Color Coding Tabs and Ergonomic Grip, Extendable Handle | |
For use on floors, walls, and ceilings in cleanroom environments. Ideal for the pull-and-lift cleaning technique |
Straight Extendable Roller Sponge Mop | |
58.5 to 76.25 in. | |
10 in. | |
Roll-O-matic Self Wringing Cleanroom Mops | |
Aluminum |