Learn More
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Labels
Label curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Brady™ handheld label maker.
$70.31 - $117.20
Specifications
Adhesive Type Permanent Acrylic
Color White
Finish Matte
Material Polyester
Print Technology Thermal Transfer
View More Specs
Products 13
Catalog Number Mfr. No. Dimensions (LxW) For Use With (Equipment) Format Price Quantity  
Catalog Number Mfr. No. Dimensions (LxW) For Use With (Equipment) Format Price Quantity  
17-012-260
View Documents Promotion Details
Brady™
M5155492
31.75 mm (W) x 6.35 mm (H) BMP51;BMP53;M511;M510 Die-cut
Each for $83.89
Only null left
17-012-117
View Documents Promotion Details
Brady™
M5130492
0.375 x 0.825 in. BMP41, BMP51, M511 Die-cut
Each for $92.51
Only null left
17-012-267
View Documents Promotion Details
Brady™
M5112492
31.75 mm (W) x 9.53 mm (H) BMP51;BMP53;M511;M510 Die-cut
Each for $106.09
Only null left
17-012-312
View Documents Promotion Details
Brady™
M583492
0.5 x 0.5 in. BMP41, BMP51, M511 Die-cut
Each for $70.31
Only null left
17-012-317
View Documents Promotion Details
Brady™
M5117492
22.86 mm (W) x 12.70 mm (H) x 11.18 mm (Dia) BMP51;BMP53;M511;M510 Die-cut
Each for $109.95
Only null left
17-012-121
View Documents Promotion Details
Brady™
M5120492
25.40 mm (W) x 12.70 mm (H) x 9.53 mm (Dia) BMP51;BMP53;M511;M510 Die-cut
Each for $105.06
Only null left
17-012-316
View Documents Promotion Details
Brady™
M5131492
0.5 x 1 in. BMP41, BMP51, M511 Die-cut
Each for $101.17
Only null left
17-012-208
View Documents Promotion Details
Brady™
M595492
41.28 mm (W) x 15.24 mm (H) x 9.53 mm (Dia) BMP51;BMP53;M511;M510 Die-cut
Each for $117.20
Only null left
17-012-302
View Documents Promotion Details
Brady™
M5156492
22.86 mm (W) x 25.40 mm (H) BMP51;BMP53;M511;M510 Die-cut
Each for $113.62
Only null left
17-012-289
View Documents Promotion Details
Brady™
M582492
0.375 x 0.375 in. BMP41, BMP51, M511 Die-cut
Each for $70.31
Only null left
17-012-213
View Documents Promotion Details
Brady™
M5125492
44.45 mm (W) x 25.40 mm (H) BMP51;BMP53;M511;M510 Die-cut
Each for $98.68
Only null left
17-012-191
View Documents Promotion Details
Brady™
M5124492
12.70 mm (W) x 41.91 mm (H) BMP51;BMP53;M511;M510 Die-cut
Each for $98.68
Only null left
17-012-318
View Documents Promotion Details
Brady™
M4C500492
25 ft. x 0.5 in. BMP41, BMP51, M511 Continuous
Each for $101.17
Only null left
Description
Description
  • Ideal for laboratory use on curved surfaces including test tubes, vials and straw pipettes
  • Withstands chemicals, abrasions and extreme temperatures, including storage in liquid nitrogen
  • Made with a permanent acrylic adhesive for tight bonding onto smooth curved surfaces such as glass and polypropylene
Specifications
Specifications
Permanent Acrylic
Matte
Thermal Transfer
Non-sterile
White
Polyester
Labels
-320°F to 230°F
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'