missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, ACS, 88%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor F109020LT

 View more versions of this product

Catalog No. 18610662


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

64-18-6,7732-18-5
88%
20 L
2e-05
1
ACS
Poly Drum

Chemical Identifiers

64-18-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.