missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid, 88%, ACS Reagent, Reagents
Formic Acid, 88%, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$212.70 - $544.20
Chemical Identifiers
CAS | 64-18-6 |
---|---|
Molecular Formula | CH2O2 |
Molecular Weight (g/mol) | 46.03 |
InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
Synonym | Methanoic Acid, Formylic Acid, Aminic Acid |
IUPAC Name | formic acid |
SMILES | OC=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-338-177
|
Labchem Inc
C110500025D1 |
2.5 L |
Each for $421.13
|
|
|||||
01-338-176
|
Labchem Inc
C11050004A |
4 L |
Each for $544.20
|
|
|||||
01-338-175
|
Labchem Inc
C1105000500A |
500 mL |
Each for $212.70
|
|
|||||
Chemical Identifiers
64-18-6 | |
46.03 | |
Methanoic Acid, Formylic Acid, Aminic Acid | |
OC=O |
CH2O2 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid |
Specifications
<2 | |
64-18-6,7732-18-5 | |
Colorless | |
Methanoic Acid, Formylic Acid, Aminic Acid | |
OC=O | |
46.03 | |
2.5 L | |
Liquid | |
Formic Acid, 88%, ACS Reagent |
8°C | |
106°C | |
CH2O2 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
ACS | |
Glass Bottle | |
1.22 |