missing translation for 'onlineSavingsMsg'
Learn More
Learn More

Formamide (Molecular Biology), Fisher BioReagents
For nucleic acid hybridization and sequencing
$280.55 - $280.55
Chemical Identifiers
CAS | 75-12-7 |
---|---|
Molecular Formula | CH3NO |
Molecular Weight (g/mol) | 45.04 |
MDL Number | MFCD00007941 |
InChI Key | ZHNUHDYFZUAESO-UHFFFAOYSA-N |
Synonym | methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 |
PubChem CID | 713 |
ChEBI | CHEBI:48431 |
IUPAC Name | formamide |
SMILES | NC=O |
Description
Molecular Biology-grade Formamide is commonly used in nucleic acid hybridization and sequencing. It requires pretreatment with a mixed-bed resin.Chemical Identifiers
75-12-7 | |
45.04 | |
ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
713 | |
formamide |
CH3NO | |
MFCD00007941 | |
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
CHEBI:48431 | |
NC=O |
Specifications
4 to 5 | |
75-12-7 | |
1.133g/cm³ | |
500 mL | |
Glass Bottle | |
≥99 % | |
H2NCH:O | |
0.08mbar at 20°C | |
15, 4265 | |
ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
formamide | |
713 | |
45.04 | |
Molecular Biology |
2°C | |
0.1 max. at 280nm | |
210°C | |
Pass Test | |
Liquid | |
CH3NO | |
MFCD00007941 | |
3.75 mPa/s at 20°C | |
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
NC=O | |
45.04 | |
CHEBI:48431 | |
≥99% | |
Formamide |
Safety and Handling

Recommended Storage : 4°C