missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Solution, min. 37% GR for Analysis, ACS, Reagent Ph Eur Grade, MilliporeSigma™
$208.59 - $4,169.83
Chemical Identifiers
CAS | 50-00-0 |
---|---|
Molecular Formula | CH2O |
Molecular Weight (g/mol) | 30.03 |
MDL Number | MFCD00003274 |
InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
Synonym | formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde |
PubChem CID | 712 |
ChEBI | CHEBI:16842 |
IUPAC Name | formaldehyde |
SMILES | C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
M1040031000
|
Technidata
1040031000 |
1 L | Plastic bottle |
N/A
|
|
||||
M1040032500
|
Technidata
1040032500 |
2.5 L | Plastic bottle |
N/A
|
|
||||
M1040035000
|
Technidata
1040035000 |
5 L | Plastic bottle |
N/A
|
|
||||
M1040039025
|
Technidata
1040039025 |
25 L | Plastic container |
N/A
|
|
||||
Description
For analysisChemical Identifiers
50-00-0 | |
30.03 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
Specifications
50-00-0,67-56-1 | |
min. 37% | |
Plastic bottle | |
MFCD00003274 | |
UN2209 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
ACS/Reagent Ph. Eur. | |
Formaldehyde solution, Stabilized with about 10% methanol |
ca.300°C | |
Stinging | |
CH2O | |
1 L | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
C=O | |
30.03 | |
CHEBI:16842 | |
Liquid |
Safety and Handling
H227: Combustible liquid.
H301 + H311 + H331: Toxic if swallowed
DOTInformation : DOT Class: 8, Packaging Group: III