missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Solution, 20% v/v, Reagents
Formaldehyde Solution, 20% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$131.20 - $243.07
Specifications
Chemical Name or Material | Formaldehyde Solution, 20% v/v |
---|---|
CAS | 7732-18-5,50-00-0 |
Color | Colorless |
Boiling Point | 100°C |
Packaging | Poly Bottle |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-335-273
|
Labchem Inc
CF1011001A |
1 L |
Each for $131.20
|
|
|||||
01-335-272
|
Labchem Inc
CF1011004A |
4 L |
Each for $243.07
|
|
|||||
Specifications
Formaldehyde Solution, 20% v/v | |
Colorless | |
Poly Bottle | |
1.03 | |
20% | |
Liquid |
7732-18-5,50-00-0 | |
100°C | |
HCHO/H2O | |
Formalin | |
Reagent |