missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde, Purified. 37%, Reagents
Formaldehyde, Purified. 37%, Technical, for general laboratory use. Manufactured in ISO 9001 facility.
$87.30 - $802.87
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | Formalin, Methanal |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-337-238
|
Labchem Inc
C22085001A |
1 L |
Each for $147.20
|
|
|||||
01-337-237
|
Labchem Inc
C220850020F |
20 L |
Each for $802.87
|
|
|||||
01-337-236
|
Labchem Inc
C22085004A |
4 L |
Each for $242.60
|
|
|||||
01-337-235
|
Labchem Inc
C2208500500A |
500 mL |
Each for $87.30
|
|
|||||
Chemical Identifiers
7732-18-5 | |
18.02 | |
Formalin, Methanal | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
7732-18-5,50-00-0,67-56-1 | |
Colorless | |
Poly Bottle | |
1 L | |
Formalin, Methanal | |
O | |
18.02 | |
Liquid |
96°C | |
96°C | |
H2O | |
1.0975 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Technical | |
Formaldehyde, Purified. 37% |