missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde, 37%, USP, Spectrum™ Chemical
FO110, 50-00-0, HCHO
Supplier: Spectrum Chemical Mfg Cor FO110200LTBL
Description
Spectrum™ Chemical Formaldehyde, 37 Percent, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
50-00-0 | |
Drum | |
MFCD00003274 | |
UN1198 | |
C=O | |
30.03 | |
USP |
100% | |
CH2O | |
200 L | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
34.5% |
Chemical Identifiers
50-00-0 | |
30.03 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
C=O |
CH2O | |
MFCD00003274 | |
formaldehyde |