missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 37%, USP, Spectrum™ Chemical

FO110, 50-00-0, HCHO

Supplier:  Spectrum Chemical Mfg Cor FO1101LT

 View more versions of this product

Catalog No. 18602831

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Formaldehyde, 37 Percent, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

50-00-0
Poly Bottle
MFCD00003274
UN1198
C=O
30.03
USP
100%
CH2O
1 L
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
34.5%

Chemical Identifiers

50-00-0
30.03
WSFSSNUMVMOOMR-UHFFFAOYSA-N
C=O
CH2O
MFCD00003274
formaldehyde
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions