missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ FoamAway™ Irradiated AOF (animal origin-free) Antifoaming Agent DFS Item

FoamAway™ Irradiated AOF (animal origin-free) Antifoaming Agent

$510.00 - $1348.00

Specifications

Classification Animal Origin-Free
Content And Storage Store at room temperature. Shake well before use. Do not filter.
Culture Type Suspension Cell Culture
For Use With (Application) Cell Culture
Product Line Gibco™
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A1036902 DFS Item
View Documents
Gibco™
A1036902
0.5 L in a 1 L bag
Each for $510.00
Only null left
 
A1036901 DFS Item
View Documents
Gibco™
A1036901
2.5 L bag
Each for $1,348.00
Only null left
 
Description

Description

FoamAway™ Irradiated AOF is a ready-to-use antifoaming agent designed to offer high performance and convenience in combating foam in your cell culture system. It is prediluted, presterilized, and packaged to connect directly to your bioreactor with no further processing required.
Specifications

Specifications

Animal Origin-Free
Suspension Cell Culture
Gibco™
No Phenol Red
Liquid
Store at room temperature. Shake well before use. Do not filter.
Cell Culture
Antifoaming Agent
Room Temperature
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.