missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Permaloc™ Formalin Containers
Double-click locking ensures specimen integrity while guarding against lost specimens through accidental spills
Supplier: Fisherbrand™ 22126353
Description
- Patented double-clicking lock design
- 1st audible“click”ensures closure
- 2nd audible“click”achieves full 95kPa certification
- Available in two convenient sizes –20mL (10mL filled) neutral-buffered formalin;60mL (30mL filled) with wide mouth (53mm) opening neutral-buffered formalin
- Leak tested and independently 95kPa certified
- Avoid employee/patient exposure to hazardous chemicals
- Improve efficiency of specimen transport to the laboratory
- Economical bulk packaging with inner convenient transport packs help grow your Outreach Programs
Specifications
Polypropylene | |
120 mL, pre-filled 60 mL | |
Permaloc™ Screw Cap | |
8 cm | |
192/Cs. |
Non-sterile | |
Polyethylene | |
None | |
6 cm | |
6 cm |