missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cryovial™ Storage Boxes

Durable, stackable polycarbonate storage boxes for vials from 1 to 5mL

Supplier:  Fisherbrand™ T314281R

 View more versions of this product

Catalog No. 361054055


Only null left
Explore available promotions

Description

Description

  • Withstand temperatures from -196° to +121°C (-321° to +250°F)
  • On 25- and 81-place boxes, a colored plastic grid insert separates the cover from the base
  • The 100-place boxes feature a colored base instead of a grid insert
  • Transparent cover allows view of box contents and is keyed to the base to prevent misalignment
  • All boxes are supplied with a vial-removal tool to facilitate removal of vials from a full box
  • Autoclavable
Specifications

Specifications

Red
81 tubes
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions