missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Ficoll-Paque™ PLUS Media
Recognized standard in laboratories worldwide for isolation of human lymphocytes from peripheral blood. Cytiva™ Ficoll-Paque™ PLUS Media maintains the viability and a representative distribution of B and T lymphocytes.
Supplier: Cytiva 17144002
Description
- Ready-to-use sterile medium
- Density media optimized for the isolation of lymphocytes from peripheral blood using simple centrifuge-based protocol
- High yield
- Maintains viability and representative distribution of B and T lymphocytes
- Low levels of endotoxin (<0.12 EU/mL)
Specifications
6 x 100 mL | |
< 0.12 EU/mL | |
Centrifugation Media | |
Sterile |
Cell isolation | |
Colorless to Slightly Yellow | |
1.078g/mL |