Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Organize vial storage within cryo boxes with these fiberboard box dividers.
Supplier: Thermo Scientific™ 820064
Description
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
- Use with fiberboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
Specifications
Each | |
64 (8 x 8 Grid) |