missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fexofenadine hydrochloride

Chemical Identifiers

CAS 153439-40-8
Molecular Formula C32H40ClNO4
Molecular Weight (g/mol) 538.13
MDL Number MFCD00865710
InChI Key RRJFVPUCXDGFJB-UHFFFAOYNA-N
IUPAC Name hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride
SMILES [H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC464970010
View Documents
Thermo Scientific™
464970010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC464972500
View Documents
Thermo Scientific™
464972500
250 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

153439-40-8
538.13
RRJFVPUCXDGFJB-UHFFFAOYNA-N
[H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
C32H40ClNO4
MFCD00865710
hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride

Specifications

153439-40-8
MFCD00865710
RRJFVPUCXDGFJB-UHFFFAOYNA-N
hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride
538.12
C32H40ClNO4
1 g
[H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
538.13
Fexofenadine hydrochloride
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only